Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Anhydrous, Purified, Reagents

Catalog No. 01337105
Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg

Potassium Fluoride, Anhydrous, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 859°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade Reagent
Packaging Fiber Drum
Specific Gravity 2.481
Chemical Name or Material Potassium Fluoride, Anhydrous, Purified
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.