Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride, Anhydrous, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605560
Click to view available options
Quantity:
25 g
125 g
1 kg
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

P1306, 7789-23-3, KF

Spectrum™ Chemical Potassium Fluoride, Anhydrous, Reagent, ACS is a source of the fluoride ion, and is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

CAS Max % 1
Quantity 1 kg
Percent Purity 99%
Grade ACS
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.