Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals

Catalog No. AA1155530
Click to view available options
Quantity:
25 g
250 g
1 kg

Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]

Specifications

Melting Point 41°C
Physical Form Crystalline
Quantity 250 g
UN Number UN1812
Merck Index 14,7632
Solubility Information Freely soluble in waterSoluble in water; Insoluble in ethanol.
IUPAC Name potassium;fluoride;dihydrate
Formula Weight 94.13 (58.10 Anhydrous)
Percent Purity ≥98.5%
Odor Odorless
Refractive Index 1.352
Sensitivity Hygroscopic
Density 2.5 g/mL
Chemical Name or Material Potassium fluoride dihydrate
Show More Show Less
Hazard Category H301+H311+H331-H318
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID
EINECSNumber 232-151-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.