Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluorosulfate, 99.5%

Catalog No. p-7051035
Click to view available options
Quantity:
10 g
50 g

CAS: 13455-22-6 | FKO3S | 138.15 g/mol

Use as primary and secondary intermediate. Finds application in chemical research..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-22-6
Molecular Formula FKO3S
Molecular Weight (g/mol) 138.15
MDL Number MFCD00216618
InChI Key VXJAYNWISQFORV-UHFFFAOYSA-M
Synonym potassium fluorosulfate, potassium sulfurofluoridate, potassium fluorosulphate, fluorosulfonyloxypotassium, acmc-1c0t1, potassium fluorosulfonyl oxidanide, potassium ion o-fluorosulfonyl oxidanidolate, fluorosulfuric acid,potassium salt 8ci,9ci
PubChem CID 23678652
IUPAC Name potassium sulfurofluoridate
SMILES [K+].[O-]S(F)(=O)=O

Specifications

Melting Point 350°C (decomposition)
Color White
Physical Form Lump/Powder
Quantity 10 g
Linear Formula KSO3F
UN Number UN3260
Solubility Information Hydrolysis.
Formula Weight 138.16
Percent Purity 99.5%
Sensitivity Moisture sensitive
Chemical Name or Material Potassium fluorosulfate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.