Alfa Aesar™ Potassium heptafluoroniobate(V), 99.5%

Manufacturer: Alfa Aesar™ 5111009

Quantity 10g

 View more versions of this product

Catalog No. AA5111009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16924-03-1
    Molecular Formula K2NbF7
    Formula Weight 304.1
    MDL Number MFCD00042368
    UN Number UN3288
    Chemical Name or Material Potassium heptafluoroniobate (V)

    Specifications

    Formula Weight 304.1
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Hygroscopic
    MDL Number MFCD00042368
    TSCA TSCA
    Physical Form Powder
    UN Number UN3288
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.
    Molecular Formula K2NbF7
    CAS 16924-03-1
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Chemical Name or Material Potassium heptafluoroniobate (V)
    Assay 99.5%

    GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.

    GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit