Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachloroosmate(IV), min 39% Os

Catalog No. p-43135
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 16871-60-6 | Cl6K2Os | 481.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16871-60-6
Molecular Formula Cl6K2Os
Molecular Weight (g/mol) 481.11
MDL Number MFCD00011371

Specifications

Melting Point 600.0°C
Color Bordeaux to Purple
Physical Form Powder
Quantity 1 g
Linear Formula K2OsCl6
Merck Index 15, 7756
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
Formula Weight 481.11
Percent Purity ≥39% (Os)
Packaging Glass bottle
Density 3.42
Chemical Name or Material Potassium hexachloroosmate(IV), min 39% Os
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 240-893-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.