Alfa Aesar™ Potassium hexachloroosmate(IV), Os 38.7% min

Manufacturer: Alfa Aesar™ 1217706

Quantity 5g

 View more versions of this product

Catalog No. AA1217706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16871-60-6
    Linear Formula K2OsCl6
    Formula Weight 481.12
    MDL Number MFCD00011371
    Merck Index 14,7635
    Chemical Name or Material Potassium hexachloroosmate(IV)

    Specifications

    Formula Weight 481.12
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Linear Formula K2OsCl6
    CAS 16871-60-6
    Merck Index 14,7635
    Color Dark red
    Quantity 5g
    Solubility Information Soluble in water; Sparingly soluble in alcohol
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Powder
    Sensitivity Hygroscopic
    MDL Number MFCD00011371
    Concentration Os 38.7% min.
    Chemical Name or Material Potassium hexachloroosmate(IV)

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit