Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachloroosmate(IV), Os 38.7% min

Catalog No. p-7048951
Click to view available options
Quantity:
0.25 g
1 g
5 g

CAS: 16871-60-6

Potassium hexachloroosmate(IV) used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16871-60-6
MDL Number MFCD00011371

Specifications

Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 0.25 g
Linear Formula K2OsCl6
Merck Index 14,7635
Solubility Information Soluble in water; Sparingly soluble in alcohol
Formula Weight 481.12
Concentration or Composition (by Analyte or Components) Os 38.7% min.
Sensitivity Hygroscopic
Chemical Name or Material Potassium hexachloroosmate(IV)
Show More Show Less
Hazard Category H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 240-893-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.