Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium hexachloroplatinate(IV), ca. 40% Pt
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 250.0°C |
| Color | Black |
| Physical Form | Powder |
| Quantity | 1 g |
| Linear Formula | K2PtCl6 |
| Merck Index | 15,7757 |
| Solubility Information | Solubility in water: 50g/L (95°C). Other solubilities: insoluble in alcohol |
| IUPAC Name | dipotassium hexachloroplatinumdiuide |
| Formula Weight | 486.01 |
| Percent Purity | ∼40% (Pt) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled. Toxic if swallowed. Causes serious eye damage. May cause an allergic skin reaction. May be corrosive to metals. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doc |
| EINECSNumber | 240-979-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction