Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 23% min

Catalog No. p-7059413
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA4454703

Supplier: Thermo Scientific Chemicals 04454703

Only null left
Add to Cart

CAS: 25443-63-4 | K3RuCl6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25443-63-4
Molecular Formula K3RuCl6
MDL Number MFCD00011390
Synonym ruthenium 6+ hexachloride, tripotassium hexachlororuthenate, ruthenium hexachloride, ruthenium chloride rucl6 , oc-6-11, ruthenate 3-, tripotassium, oc-6-11

Specifications

Physical Form Crystalline
Quantity 1 g
Formula Weight 431.09
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Ru 23% min
Assay (metals basis)
Chemical Name or Material Potassium hexachlororuthenate (III)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-983-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.