Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexafluorophosphate, 98%

Catalog No. AAA1208030
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 17084-13-8 | F6KP | 184.062 g/mol

Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17084-13-8
Molecular Formula F6KP
Molecular Weight (g/mol) 184.062
MDL Number MFCD00011412
InChI Key YZDGRYDIGCWVND-UHFFFAOYSA-N
Synonym potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
PubChem CID 23688904
IUPAC Name potassium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[K+]

Specifications

Melting Point 575°C
pH 3.0 to 7.0
Quantity 250 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 184.07
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.55
Chemical Name or Material Potassium hexafluorophosphate
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 241-143-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.