Learn More
Potassium hexafluorophosphate, 98%
Description
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and oxidizing agents.
Specifications
Specifications
| Melting Point | 575°C |
| pH | 3.0 to 7.0 |
| Quantity | 50 g |
| UN Number | UN3260 |
| Solubility Information | Soluble in water. |
| Formula Weight | 184.07 |
| Percent Purity | 98% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 2.55 |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H314 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 241-143-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.