Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexafluorozirconate, 99%, Thermo Scientific Chemicals

Catalog No. p-7080353
Click to view available options
Quantity:
500 g
2 kg

For manufacture of metallic zirconium

Employed in the manufacture of metallic zirconium. Used in the production of aluminum magnesium alloy, ceramics and glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16923-95-8
Molecular Formula F6K2Zr
Molecular Weight (g/mol) 283.411
MDL Number MFCD00042547
InChI Key BJZIJOLEWHWTJO-UHFFFAOYSA-H
Synonym potassium hexafluorozirconate, zirconium potassium fluoride, potassium fluorozirconate, f6zr.2k, dipotassium hexafluorozirconate, potassium hexafluorozirconate iv, dipotassium hexafluorozirconiumdiuide, dipotassium hexafluorozirconate iv
PubChem CID 56828182
IUPAC Name dipotassium;hexafluorozirconium(2-)
SMILES F[Zr-2](F)(F)(F)(F)F.[K+].[K+]

Specifications

Quantity 500 g
UN Number UN3288
Merck Index 14,7639
Solubility Information Slightly soluble in cold water, soluble in hot water
Formula Weight 283.41
Percent Purity 99%
Odor Odorless
Density 3.48 g/mL
Chemical Name or Material Potassium hexafluorozirconate
Hazard Category H301-H318
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280-P301+P310-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 240-985-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.