Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexahydroxyplatinate(IV), Premion™, 99.95% (metals basis), Pt 51.5% min

Catalog No. AA4101406
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA4101406

Supplier: Thermo Scientific Chemicals 04101406

Only null left
Add to Cart

CAS: 12285-90-4 | H12K2O6Pt+2 | 381.371 g/mol

Potassium hexahydroxyplatinate(IV) is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12285-90-4
Molecular Formula H12K2O6Pt+2
Molecular Weight (g/mol) 381.371
MDL Number MFCD00064671
InChI Key LTROBPQZKJGHPB-UHFFFAOYSA-N
Synonym potassium hexahydroxoplatinate, dipotassium platinate, potassium hexahydroxyplatinate iv, platinum potassium hydroxide, h6k2o6pt, potassium hexahydroxoplatinate iv, potassium hexahydroxyplatinum iv, potassium hexahydroxyplatinate, dipotassium hexahydroxyplatinumdiuide, premion metalsbasis ,ptmin
PubChem CID 16717651
IUPAC Name dipotassium;platinum;hexahydrate
SMILES O.O.O.O.O.O.[K+].[K+].[Pt]

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 375.34
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 51.5% min.
Assay (metals basis)
Chemical Name or Material Potassium hexahydroxyplatinate(IV)
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 235-554-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.