Learn More
Potassium hydrogen carbonate, 99%
Description
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek′s agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong acids.

Specifications
Specifications
Quantity | 10,000 g |
Merck Index | 14,7609 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 100.12 |
Percent Purity | 99% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.17 |
Chemical Name or Material | Potassium hydrogen carbonate |
Safety and Handling
EINECSNumber | 206-059-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.