Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, 97%

Catalog No. AC457931000
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. AC457931000

Supplier: Thermo Scientific Chemicals 457931000

Only null left
Add to Cart

CAS: 194491-32-2 | C10H12MgN2Na2O8 | 358.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194491-32-2
MDL Number MFCD00078217
InChI Key AWNVVAMWLMUZOZ-UHFFFAOYSA-J
IUPAC Name magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Assay Percent Range 96.0 % min. (on dry substance)
Molecular Formula C10H12MgN2Na2O8
Quantity 100 g
Solubility Information Solubility aq. soln.: soluble.
Molecular Weight (g/mol) 358.50
Formula Weight 358.5
Chemical Name or Material Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.