Learn More
Potassium hydrogen L-tartrate, 98+%
Description
Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.
Solubility
Soluble in water and dilute mineral acid. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼267°C (decomposition) |
| Density | 1.954 |
| Quantity | 5000 g |
| Beilstein | 6119985 |
| Merck Index | 14,7615 |
| Solubility Information | Soluble in water and dilute mineral acid. Insoluble in alcohol. |
| PubChem CID | 376198361 |
| Formula Weight | 188.18 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Potassium hydrogen L-tartrate |
Safety and Handling
| EINECSNumber | 212-769-1 |
| RTECSNumber | WW8223000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.