Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, anhydrous, 99.9%, ACS Grade, DriSolv, MilliporeSigma™

Catalog No. p-4382912
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Septum-sealed Glass Bottle

Chemical Identifiers

Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8 (200g/L, H2O, 20°C)
Melting Point -108.3°C
CAS 109-99-9
Color Colorless
Appearance Clear liquid, Free from particulates
Density 0.89g/cm3 at 20°C
Boiling Point 66.1°C
Quantity 100 mL
Flash Point 15.15°C (4.7°F), Closed cup
Ignition Point 320.85°C (609.5°F)
Odor Ether-like
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.9% min.
Linear Formula CH2(CH2)3O
Vapor Pressure 173hPa (20°C)
UN Number UN 2056
Solubility Information Soluble in water
Formula Weight 72.11
Grade ACS
Chemical Name or Material Tetrahydrofuran, DriSolv™
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-726-8
RTECSNumber LU5950000
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.