Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydrogen phthalate, 99+%

Catalog No. ac417955000
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 295.0°C to 300.0°C
Color Red
pH 4 to 4.02 (0.05M soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula 2-(HO2C)C6H4CO2K
Quantity 500 g
Beilstein 09, 791
Merck Index 15, 7733
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 330g/L water (100°C), slightly soluble in alcohol
Formula Weight 204.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Potassium hydrogen phthalate, 99+%
Show More Show Less
EINECSNumber 212-889-4
RTECSNumber CZ4326000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.