Learn More
Potassium hydrogen sulfate, 97%
Description
Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.
Solubility
Soluble in water, acetone and ethanol.
Notes
Hygroscopic. Incompatible with strong bases, strong oxidizing agents and alcohols.
Specifications
Specifications
| Melting Point | 195°C to 197°C (decomposition) |
| Quantity | 2500 g |
| Linear Formula | KHSO4 |
| UN Number | UN2509 |
| Merck Index | 14,7613 |
| Solubility Information | Soluble in water,acetone and ethanol. |
| Formula Weight | 136.17 |
| Percent Purity | 97% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H335 Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROGEN SULFATE |
| EINECSNumber | 231-594-1 |
| RTECSNumber | TS7200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.