Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydrogen sulfate, fused, ACS, mixture of potassium pyrosulfate, K{2}S{2}O{7}, and potassium bisulfate, KHSO{4}

Catalog No. AA1380022
Click to view available options
Quantity:
100 g
500 g

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It is also used as a disintegrating agent in analytical chemistry. Further, it is used to make fertilizers and in the preparation of potassium persulfate. In addition to this, it is used as a catalyst in organic preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00066385

Specifications

Melting Point 214°C
Color White
Physical Form Fused Powder
Quantity 100 g
Solubility Information Soluble in water,isopropyl alcohol and acetone.
Formula Weight 136.17
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White fused powder
Chemical Name or Material Potassium hydrogen sulfate, fused
Show More Show Less
Hazard Category H314-H331
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8, Packing Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.