Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.0100 N (N/100) in Methanol, Ricca Chemical
SDP

Catalog No. R6264000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R6264000500 500 mL Natural Poly Bottle
R62640001A 1 L Natural Poly Bottle
R62640004F 4 L Cubitainer
R62640004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R6264000500 Supplier Ricca Chemical Company Supplier No. 626416
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 67-56-1 | HKO | 56.11 g/mol

Chemical Identifiers

CAS 67-56-1 , 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Assay Percent Range 0.01000 ± 0.00005 N at 20°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Solubility in Water: Infinite
Quantity 500 mL
Grade Laboratory
Odor Characteristic alcohol
Packaging Natural Poly Bottle
Solvent or Matrix Nonaqueous Solution
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.0100 Normal (N/100) in Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.