Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.100 N (N/10) in Denatured Alcohol, Ricca Chemical
SDP

Catalog No. 6264532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
62645-32 1 L Natural Poly Bottle
6264-516 500 mL Natural Poly Bottle
62645-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 62645-32 Supplier Ricca Chemical Company Supplier No. 6264.532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64-17-5 | HKO | 56.11 g/mol

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 1.59, 96.0, 0.03
CAS Max % 1.65, 100.0, 0.03
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.100 Normal (N/10) in Denatured Alcohol
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.