Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.100 Normal (N/10) in Ethanol, Reagents

Catalog No. 01334330
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-330 4 L
01-334-331 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-330 Supplier Reagents Holdings Llc Supplier No. CP1168004C
Add to Cart
Add to Cart

Potassium Hydroxide, 0.100 Normal (N/10) in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name ethanol
SMILES CCO

Specifications

pH 13
Color Colorless
Boiling Point 78°C
Quantity 4 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.82
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.100 Normal (N/10) in Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.