Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical
SDP

Catalog No. R62540001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R62540001A 1 L Natural Poly Bottle
R6254000500 500 mL Natural Poly Bottle
R62540004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R62540001A Supplier Ricca Chemical Company Supplier No. R62540001A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0 , 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Assay Percent Range 0.2000 ± 0.0005 N at 20°C
Solubility Information Solubility in Water: Infinite
Quantity 1 L
Grade Laboratory
Odor Characteristic alcohol odor
Packaging Natural Poly Bottle
Solvent or Matrix Nonaqueous Solution
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.2N in Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.