Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol), Reagents

Catalog No. 01334466
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-334-466 4 L Poly Bottle
01-334-469 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-466 Supplier Reagents Holdings Llc Supplier No. CP1014004A
Add to Cart
Add to Cart

Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name ethanol
SMILES CCO

Specifications

pH >13
Color Colorless
Boiling Point 78°C
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8168
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.