Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.500 N (N/2) in Isopropyl Alcohol, Ricca Chemical
SDP

Catalog No. 625751 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 67-63-0, 1310-58-3, 107-15-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 3.89, 94.0, 0.03
CAS Max % 4.05, 98.0, 0.03
Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.500 Normal (N/2) in Isopropyl Alcohol
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
ShelfLife 12 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.