Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydroxide, 10.0 N, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
1 L
4 L
30 gal.
Packaging:
Natural Poly Bottle
Poly Drum
Specifications
Specifications
Color | Colorless |
CAS Min % | 60.0, 38.44 |
CAS Max % | 62.0, 40.01 |
Grade | Laboratory |
Packaging | Poly Drum |
pH | >13 |
Quantity | 30 gal. |
Physical Form | Liquid |
Concentration or Composition Notes | 10.0N |
Chemical Name or Material | Potassium Hydroxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction