Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 10% (w/v), Ricca Chemical
SDP

Catalog No. R6130000250 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R6130000250 250 mL Natural Poly Bottle
6130-4 120 mL Natural Poly Bottle
6130-16 500 mL Natural Poly Bottle
6130-32 1 L Natural Poly Bottle
6130-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R6130000250 Supplier Ricca Chemical Company Supplier No. R6130000250A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 82.0, 15.92
CAS Max % 85.0, 16.56
Synonym potassium hydroxide
Quantity 250 mL
Percent Purity 9.94 to 10.06% w/v
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 10% (w/v)
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.