Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydroxide, 30% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
| CAS Min % | 75.0, 22.63 |
| CAS Max % | 78.0, 23.55 |
| Quantity | 4 L |
| Percent Purity | 29.94 to 30.06% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 30% (w/v) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Potassium Hydroxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction