Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCP05003A5 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
3 kg
25 kg
1000 gal.
Packaging:
Fiber Drum
Natural Poly Bottle
White Poly Wide Mouth Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCP05003A5 3 kg Natural Poly Bottle
RDCP0500500 500 g White Poly Wide Mouth Bottle
RDCP0500100 1000 gal. White Poly Wide Mouth Bottle
RDCP050025F 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RDCP05003A5 Supplier Ricca Chemical Company Supplier No. RDCP05003A5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Synonym potassium hydroxide
Quantity 3 kg
Percent Purity ≥85%
Grade ACS
Packaging Natural Poly Bottle
Physical Form Deliquescent Pellets
Chemical Name or Material Potassium Hydroxide
ShelfLife 120 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.