Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, ca. 85%, for analysis, pellets, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
10kg
25kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
1310-58-3
HKO
56.105
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3
HKO
56.105
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
Quantity 250g
Assay Percent Range ca. 85%
Linear Formula KOH
Fieser 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
Merck Index 15, 7761
Formula Weight 56.11
Physical Form Pellet
Percent Purity ca. 85%
Grade Analysis
Packaging Plastic bottle
Solubility Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether
Melting Point 360.0°C
Boiling Point 1320.0°C
Chemical Name or Material Potassium hydroxide, ca. 85 %
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger