Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Pellets, FCC, 85-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605596 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-596 6 x 500 g Poly Bottle
18-605-595 500 g Poly Bottle
18-605-597 2.5 kg Poly Bottle
18-605-598 4 x 2.5 kg Poly Bottle
18-605-599 12 kg Poly Pail
18-605-600 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-596 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1316500GMCS6
Add to Cart
Add to Cart

P1316, 1310-58-3, KOH

Spectrum™ Chemical Potassium Hydroxide, Pellets, FCC is used as a rinse or chemical peel for fruits and vegetables, a stabilizer to prolong shelf-life, and as a thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

CAS Max % 1
Quantity 6 x 500 g
Percent Purity 85 to 100.5%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.