Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution, 0.5N (N/2) (In Methanol/Certified), Fisher Chemical™

Catalog No. SP22220 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP222-20 20 L PolyPac™
SP222-1 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP222-20
  • $1,056.00
    $897.65 / Each of 1

    Save $158.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Titrated at 25°C to pH 8.2 (phenolphthalein endpoint) against standardized HCl

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3 , 67-56-1 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Boiling Point 65.6°C
Melting Point -100°C
Concentration 0.497 to 0.500N
UN Number 2924
Density 0.8g/cm³
Quantity 20 L
Formula Weight 56.11
Grade Certified
Concentration or Composition (by Analyte or Components) 0.5N (N/2)
Identification Pass Test
Packaging PolyPac™
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 0.5N (N/2), in methanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.