Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodate, 99.5%, for analysis

Catalog No. AC201771000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 7758-05-6 | IKO3 | 214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-05-6, 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

Melting Point 560°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula KIO3
Merck Index 15, 7763
Solubility Information Solubility in water: soluble
Formula Weight 214
Percent Purity 99.50%
Grade Analytical
Loss on Drying 0.5% max.
Packaging Plastic bottle
Specific Gravity 3.93
Density 3.9300g/mL
Chemical Name or Material Potassium iodate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-831-9
RTECSNumber NN1350000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.