Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate Concentrate (Certified), Fisher Chemical

Catalog No. sp233 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL

Each bottle contains exact amount of 1N concentrate required to make 1 liter of 0.1N (N/10) solution

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-05-6, 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Color Colorless
Boiling Point 100°C
Melting Point 0°C
Concentration 0.0995 to 0.1010N
Density 1.0g/cm³
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 17.5mmHg
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Potassium Iodate Concentrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.