Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Iodate-Iodide, 0.0079 Normal Standardized, Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Specifications
Chemical Name or Material | Potassium Iodate-Iodide, 0.0079 Normal Standardized |
Boiling Point | 100°C |
CAS | 7732-18-5, 7681-11-0 |
Color | Colorless |
Physical Form | Liquid |
Quantity | 4 L |
Molecular Formula | KIO3/KI/H2O |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.0021 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction