Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granules, Fisher BioReagents™ PROMO

Catalog No. BP367500S24 Shop All Fisherbrand Products
Click to view available options
:
Potassium Iodide, Granules; Packaging: Poly Bottle; Quantity: 500 g

Synonym: Thyro-Block, Kali Iodide, Thyroshield

  • Potassium salt used in buffers for molecular biology and cell culture applications
  • White, free-flowing granules
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 680°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99%
Merck Index 15, 7764
Formula Weight 166
Percent Purity ≥99%
Identification Pass Test
Loss on Drying 0.2%
Packaging Poly Bottle
Vapor Pressure 1 mmHg at 745°C
Chemical Name or Material Potassium Iodide
Show More Show Less
Hazard Statement May cause skin, eye, and respiratory tract irritation.
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.