Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodide, 99.995% (metals basis)

Catalog No. AA1097822
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7681-11-0 | IK | 166.003 g/mol

Potassium iodide is used with silver nitrate to make silver iodide an important chemical in film photography. Medicinally it is used for thyroid blockade, thyroid storm and also as an expectorant. It is a component in some hair treatment chemicals. It is also used as a fluorescence quenching agent in biomedical research, an application that takes advantage of collisional quenching of fluorescent substances by the iodide ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 681°C
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.995% (metals basis)
Solubility Information Soluble in water,alcohol,methanol,acetone and ethanol.
Formula Weight 166.01
Odor Odorless
Refractive Index 1.677
Sensitivity Hygroscopic
Appearance White crystalline powder
Vapor Pressure 1.33 hPa (1mm Hg) at 745°C
Density 3.12
Chemical Name or Material Potassium iodide
Show More Show Less
Hazard Category H372
Precautionary Statement P260-P264b-P270-P314-P501c
EINECSNumber 213-659-4
RTECSNumber TT2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.