Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodide, 99.995%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
7681-11-0
IK
166
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
7681-11-0
IK
166
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
For ultra trace analysis
Assay Percent Range 99.995%
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Physical Form Powder or Cyrstals
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Total Trace Metal Impurities 0.005% max.
Boiling Point 1330.0°C
Color White
Melting Point 680.0°C
Quantity 250g
Chemical Name or Material Potassium iodide
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 231-659-4