Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodide, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™

Catalog No. 60047493 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1,323°C
Melting Point 681°C (literature)
CAS Max % 100.0000%
Physical Form Anhydrous
Quantity 500 g
Linear Formula KI
Formula Weight 166
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 3.12 g/cm3
Chemical Name or Material Potassium iodide, Free-flowing, ACS reagent, anhydrous, ≥99%
Show More Show Less
ShelfLife 720 days from date of manufacture
EINECSNumber 231-659-4
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.