Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Iodide TS, 16.5% (w/v), Stabilized, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
Specifications
Specifications
| Color | Colorless to Yellow |
| CAS Min % | 13.89, 84.0, 0.01 |
| CAS Max % | 14.45, 88.0, 0.01 |
| Percent Purity | 16.5% |
| Grade | Laboratory |
| Packaging | Amber Poly Bottle |
| Treatment(s) | Stabilized |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction