Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium metabisulfite, 95+%

Catalog No. AA43908A3
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 16731-55-8 | K2O5S2 | 222.31 g/mol

It serves as a component of certain photographic developers and its solution is used in photographic fixing, in the textile industry for dyeing and cotton printing. It is utilized to precipitate gold from solution in aqua regia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

Melting Point 190°C (decomposition)
Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range >99%
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 222.32
Odor Pungent
Sensitivity Air Sensitive
Chemical Name or Material Potassium metabisulfite
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 240-795-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.