Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Metabisulfite, Crystal, NF, 51.8-57.6%, Spectrum™ Chemical
SDP

Catalog No. 18605645 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-605-645

Supplier: Spectrum Chemical Manufacturing Corporation P1346500GM

Only null left
Add to Cart
Add to Cart

P1346, 16731-55-8, K2S2O5

Spectrum™ Chemical Potassium Metabisulfite, Crystal, NF is used primarily as an antioxidant in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 51.8 to 57.6%
Grade NF
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.