Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Metabisulfite (Granular/NF/FCC), Fisher Chemical™

Catalog No. p-33072 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

Melting Point 150°C
Color White
pH 3.5 to 4.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 222.33
Percent Purity 51.8 to 57.5% (NF)
Grade FCC/NF
Identification Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Packaging Poly Bottle
Chemical Name or Material Potassium Metabisulfite
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.