Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium metavanadate, 99.9% (metals basis)

Catalog No. p-7050803
Click to view available options
Quantity:
25 g
100 g

CAS: 13769-43-2 | KO3V | 138.04 g/mol

Employed as corrosion inhibitors and anti-scaling agents. Used widely in chemical fertilizer industries, glass and ceramic industries, pharmaceutical industries. Also used in the process of pigment production for plastics, enamels, ceramics. Used for industrial gas desulphuration and H2S adsorption and elimination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13769-43-2
Molecular Formula KO3V
Molecular Weight (g/mol) 138.04
MDL Number MFCD00050038
InChI Key BQFYGYJPBUKISI-UHFFFAOYSA-N
Synonym potassium metavanadate, potassium vanadate, potassium m-vanadate, potassium trioxovanadate, potassium vanadium trioxide, unii-0s0516d6vi, vanadate vo31-, potassium, vanadic acid, potassium salt, potassium vanadate v kvo3 6ci,7ci, potassium dioxidovanadium 1+
PubChem CID 4571336
IUPAC Name potassium;oxido(dioxo)vanadium
SMILES [K+].[O-][V](=O)=O

Specifications

Melting Point 520°C
Physical Form Powder
Quantity 25 g
UN Number UN2864
Solubility Information Soluble in water.
Formula Weight 138.04
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Potassium metavanadate
Hazard Category H302+H332-H319-H361d-H372
Hazard Statement GHS H Statement
H330-H302-H315-H319-H335
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: POTASSIUM METAVANADATE
EINECSNumber 237-388-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.