Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium methoxide, pure, 0.1N solution in toluene/methanol, AcroSeal™

Catalog No. ac429101000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 865-33-8 | CH3KO | 70.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-33-8, 67-56-1, 108-88-3
Molecular Formula CH3KO
Molecular Weight (g/mol) 70.13
MDL Number MFCD00012416
InChI Key BDAWXSQJJCIFIK-UHFFFAOYSA-N
Synonym potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
PubChem CID 23664618
IUPAC Name potassium;methanolate
SMILES C[O-].[K+]

Specifications

Quantity 100 mL
Linear Formula CH3OK
Flash Point 7°C
Solubility Information Solubility in water: may decompose
Formula Weight 70.13
Grade Pure
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.85
Density 0.8500g/mL
Chemical Name or Material Potassium methoxide, Pure, 0.1N solution in toluene/methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Toxic if swallowed. Causes skin irritation. May cause drowsiness or dizziness. Toxic if inhaled. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
EINECSNumber 212-736-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.