Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium molybdate, 95%

Catalog No. p-4440153
Click to view available options
Quantity:
100 g
500 g

CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-49-6
Molecular Formula K2MoO4
Molecular Weight (g/mol) 238.14
MDL Number MFCD00011370
InChI Key NUYGZETUIUXJCO-UHFFFAOYSA-N
Synonym potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
PubChem CID 6093816
IUPAC Name dipotassium;dioxido(dioxo)molybdenum
SMILES [K+].[K+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 919.0°C
Quantity 100 g
Assay Percent Range 94% min
Linear Formula K2MoO4
Merck Index 15,7768
Formula Weight 238.14
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Potassium molybdate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 236-599-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.