Learn More
Potassium nitrate, 99.994% (metals basis)
Description
Potassium nitrate has several applications. It is used as an oxidizer, food preservative, fertilizer, and an active ingredient of condensed aerosol fire suppression systems. It is a powerful nitrating agent in sulfuric acid medium. In addition, it also used as an electrolyte in a salt bridge and thermal storage medium in power generation systems. Ternary salts containing potassium nitrate have been reported to improve the heat storage capacity in the molten salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 400°C (decomposition) |
Melting Point | 334°C |
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
UN Number | UN1486 |
Merck Index | 14,7648 |
Solubility Information | Freely soluble in water. Slightly soluble in ethanol. |
IUPAC Name | potassium;nitrate |
Formula Weight | 101.11 |
Show More |
Safety and Handling
Hazard Category | H272 |
Hazard Statement | GHS H Statement H272 May intensify fire oxidizer. |
Precautionary Statement | P210-P220-P221-P280-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM NITRATE |
EINECSNumber | 231-818-8 |
RTECSNumber | TT3700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.