Learn More
Potassium nitrate, 99.999%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 400.0°C |
| Melting Point | 334.0°C |
| Color | White |
| Physical Form | Crystals and/or Chunks |
| Quantity | 50 g |
| Assay Percent Range | 99.999% |
| Linear Formula | KNO3 |
| Fieser | 05,561 |
| Merck Index | 15, 7769 |
| Solubility Information | Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: May intensify fire; oxidizer. |
| Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 231-818-8 |
| RTECSNumber | TT3700000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.